missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGS9 Monoclonal Antibody (D7)
GREENER_CHOICE

Product Code. 11560372
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11560372 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11560372 Supplier Invitrogen™ Supplier No. MA1723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-723 detects RGS9-1 protein from bovine and mouse samples. MA1-723 has been successfully used in Western blot, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a ~57 kDa protein representing RGS-1 protein from bovine retinal extracts. The MA1-723 immunogen is full length, bacterially expressed RGS9-1 protein.

The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS9
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene RGS9
Gene Accession No. O46469, O54828
Gene Alias GTPase accelerating protein for heterotrimeric G-proteins; MGC111763; MGC26458; PERRS; regulator of G protein signaling 9; regulator of G protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signalling 9; Rgs9; RGS9-1; Rgs9-2; RGS9L; ROS GAP
Gene Symbols RGS9
Host Species Mouse
Immunogen Full length, bacterially expressed RGS9-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19739, 281453
Target Species Mouse, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.