missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHAMM/CD168 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76538-0.025mg
This item is not returnable.
View return policy
Description
RHAMM/CD168 Polyclonal specifically detects RHAMM/CD168 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RHAMM/CD168 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3161 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunofluorescence 5 μg/mL | |
| NP_036617 | |
| HMMR | |
| Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of human RHAMM. The immunogen is located within amino acids 80 - 130 of RHAMM. Amino Acid Squence: ERGAQDRRIQDLETELEK | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction