Learn More
Invitrogen™ RHAMM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588546
Description
Immunogen sequence: MSFPKAPLKR FNDPSGCAPS PGAYDVKTLE VLKGPVSFQK SQRFKQQKES KQNLNVDKDT TLPASARKVK SSESKKESQK NDKDLKILEK EIRVLLQERG AQDRRIQDLE TELEKMEARL NAALREKTSL SANNATLEKQ LIELTRTNEL LKSKFSENGN QKNLRILSLE LMKLRNKRET KMRGMMAKQE GMEMKLQVTQ RSLEESQGKI AQLEGKLVSI EKEKIDEKSE TEKLLEYIEE ISCASDQVEK YKLDIAQLEE NLKEKNDEIL SLKQSLEENI VILSKQVEDL NVKCQLLEKE; Positive Samples: SW480, HeLa, Jurkat, ES-2; Cellular Location: Cell surface, Cytoplasm.
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
| RHAMM | |
| Polyclonal | |
| Unconjugated | |
| HMMR | |
| AA386826; CD168; Hmmr; Hyaluronan mediated motility receptor; hyaluronan mediated motility receptor (RHAMM); hyaluronan-mediated motility receptor; hyaluronan-mediated motility receptor (RHAMM); IHABP; Intracellular hyaluronic acid-binding protein; receptor for hyaluronan-mediated motility; RHAMM | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75330 | |
| HMMR | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.