missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDD1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76248JF549
This item is not returnable.
View return policy
Description
RHBDD1 Polyclonal antibody specifically detects RHBDD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| RHBDD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| DKFZp547E052, EC 3.4.21, EC 3.4.21.-, MGC117258, rhomboid domain containing 1, rhomboid domain-containing protein 1 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human RHBDD1. The immunogen is located within the last 50 amino acids of RHBDD1. Amino Acid Squence: WDRGNTRNSPPPYG | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 84236 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction