missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDD2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76249R
This item is not returnable.
View return policy
Description
RHBDD2 Polyclonal antibody specifically detects RHBDD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| RHBDD2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 550 | |
| H_RG122E10.2a, H_RG122E10.2b, rhomboid domain containing 2, rhomboid domain-containing protein 2, rhomboid, veinlet-like 7, veinlet-like 7 (Drosophila) | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the center of human RHBDD2. The immunogen is located within amino acids 220 - 270 of RHBDD2. Amino Acid Squence: AERRAAQSRKLMPVPGSY | |
| 0.1 mL | |
| Apoptosis | |
| 57414 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction