missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDD3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76250AF405
This item is not returnable.
View return policy
Description
RHBDD3 Polyclonal antibody specifically detects RHBDD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| RHBDD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 405 | |
| C22orf3, chromosome 22 open reading frame 3, HS984G1A, pituitary tumor apoptosis, PTAG, rhomboid domain containing 3, rhomboid domain-containing protein 3 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human RHBDD3. The immunogen is located within amino acids 170 - 220 of RHBDD3. Amino Acid Squence: FRWLEPSERRLQV | |
| 0.1 mL | |
| Apoptosis | |
| 25807 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction