missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHBDF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121461
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Regulates ADAM17 protease, a sheddase of the epidermal growth factor (EGF) receptor ligands and TNF, thereby plays a role in sleep, cell survival, proliferation, migration and inflammation. Does not exhibit any protease activity on its own. [UniProt].
Specifications
| RHBDF2 | |
| Polyclonal | |
| Unconjugated | |
| RHBDF2 | |
| Inactive rhomboid protein 2; IRHOM2; RHBDF2; RHBDL5; RHBDL6; rhomboid 5 homolog 2; Rhomboid family member 2; rhomboid veinlet-like protein 5; Rhomboid veinlet-like protein 6; TEC; TOC; TOCG | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79651 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 3.65 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6PJF5 | |
| RHBDF2 | |
| Recombinant Human Inactive rhomboid protein 2 protein (1-200aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction