missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93706-0.1ml
This item is not returnable.
View return policy
Description
RHBDL1 Polyclonal antibody specifically detects RHBDL1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RHBDL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.4.21.105, RHBDLrhomboid, drosophila, homolog of, rhomboid, veinlet-like 1 (Drosophila), Rhomboid-like protein 1, rhomboid-related protein 1, RRPrhomboid (veinlet, Drosophila)-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RHBDL1 (O75783). MGRVEDGGTTEELEDWDPGTSALPAPGIKQGPREQTGTGPLSQKCWEPEPDAPSQPGPALWSRGRARTQALAGGSSLQQLDPENTGFIGADTFTGLVHSHELPLDPAKLDMLVALAQSNEQGQVCYQELVDLISSKRSSSFKRAIANGQRALPRDGPLDEPGLGVYKRFVRYVAYEILPCEVDRRWYFYRHRSCPPPVFM | |
| 0.1 mL | |
| Cell Biology | |
| 9028 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction