missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHBDL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15505842
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505842 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15505842 Supplier Invitrogen™ Supplier No. PA543021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ELEEEEKMRE DGGGKDRAKS KKVHRIVSKW MLPEKSRGTY LERANCFPPP Sequence homology: Human: 100%.

RHBDL2 is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. RHBDL2 is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RHBDL2
Gene Accession No. Q9NX52
Gene Alias 9130416B15; CTF; NTF; RHBDL2; rhomboid (veinlet, Drosophila)-like 2; rhomboid like 2; rhomboid protease 2; rhomboid, veinlet-like 2; rhomboid, veinlet-like 2 (Drosophila); rhomboid-like protein 2; Rhomboid-related protein 2; Rhomboid-related protein 2, C-terminal fragment; Rhomboid-related protein 2, N-terminal fragment; RRP2
Gene Symbols RHBDL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RHBDL2 (aa 23-72).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54933
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.