missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHD Antibody (roledumab), Alexa Fluor™ 594, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27969AF594
This item is not returnable.
View return policy
Description
RHD Monoclonal antibody specifically detects RHD in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| RHD | |
| Monoclonal | |
| Alexa Fluor 594 | |
| blood group Rh(D) polypeptide, CD240D, DIIIc, RH, Rh blood group, D antigen, RH30, Rh4, RHCED, RhDCw, RHDel, RHDVA(TT), RhII, RhK562-II, RhPI, RHPII, RHXIII | |
| RHD/ CD240d | |
| 0.1 mL | |
| Cardiovascular Biology, CD Markers, Immunology | |
| 6007 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| roledumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction