missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rheb Antibody (Rheb 5D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 351.00
Specifications
| Antigen | Rheb |
|---|---|
| Clone | Rheb 5D5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689898
|
Novus Biologicals
NBP2-50273-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664377
|
Novus Biologicals
NBP2-50273 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rheb Monoclonal antibody specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Rheb | |
| 1 mg/mL | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, mTOR Pathway, Signal Transduction | |
| PBS | |
| 6009 | |
| IgG2a | |
| Protein A purified |
| Rheb 5D5 | |
| Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
| Human Rheb GST fusion | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title