missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHEB Monoclonal Antibody (GT39810)

Product Code. 16362284
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16362284 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16362284 Supplier Invitrogen™ Supplier No. MA527777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: SK-N-SH Predicted Reactivity: Mouse (98%), Rat (99%), Zebrafish (90%), Xenopus laevis (91%), Pig (99%), Sheep (99%), Rhesus Monkey (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Rheb encodes a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. This protein is vital in regulation of growth and cell cycle progression due to its role in the insulin/TOR/S6K signaling pathway. The protein has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form, and farnesylation of the protein is required for this activity. Three pseudogenes have been mapped, two on chromosome 10 and one on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHEB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT39810
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RHEB
Gene Accession No. Q15382, Q62639, Q921J2
Gene Alias GTP-binding protein Rheb; Ras homolog enriched in brain; Ras homolog enriched in brain 2; RAS-homolog enriched in brain; RHEB; Rheb1; RHEB2
Gene Symbols RHEB
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human Rheb. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19744, 26954, 6009
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.