missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoA Antibody (1B3-4A10), Novus Biologicals™
Mouse Monoclonal Antibody
€ 200.00
Specifications
| Antigen | RhoA |
|---|---|
| Clone | 1B3-4A10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:1500, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:50 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Description
RhoA Monoclonal specifically detects RhoA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| RhoA | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| RHOA | |
| IgG | |
| Protein A purified |
| 1B3-4A10 | |
| Western Blot 1:500 - 1:1500, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:50 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 387 | |
| Full-length human recombinant RhoA protein. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title