missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoA, p Ser188 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23372
This item is not returnable.
View return policy
Description
RhoA Polyclonal antibody specifically detects RhoA in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RhoA | |
| Polyclonal | |
| Western Blot 1:1000 | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| Phospho-RhoA (Ser-188) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 188 of human RhoA. This peptide sequence is highly conserved in rat, mouse, and chicken RhoA, but is not found in RhoB or RhoC proteins. [UniProt# P06749] | |
| 0.1 mL | |
| Cancer, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 387 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/mL BSA and 50% Glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction