missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOA, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB7905.100uL
This item is not returnable.
View return policy
Specifications
| RHOA | |
| Polyclonal | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| ras homolog gene family, member A | |
| RHOA | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S188 of human RHOA. | |
| RUO | |
| 387 | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| Unconjugated | |
| In PBS (50% glycerol, 1 mg/mL BSA, 0.05% sodium azide) | |
| ARH12/ARHA/RHO12/RHOH12 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This peptide sequence is highly conserved in rat and mouse RhoA, but is not found in RhoB or RhoC proteins. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction