missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RhoB VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1319)

Product Code. 30131912
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131912 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131912 Supplier Invitrogen™ Supplier No. MA554681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpaca Recombinant Monoclonal Antibody

RhoB VHH-8His-Cys-tag Recombinant Monoclonal Antibody for SPR, ICC/IF, IP

Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoB VHH-8His-Cys-tag
Applications Immunoprecipitation, Immunocytochemistry, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone SAA1319
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RHOB
Gene Accession No. P62745
Gene Alias AA017882; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; Arh6; Arhb; h6; MST081; MSTP081; oncogene RHO H6; ras homolog B; ras homolog family member B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rhoB; RHOH6; Rho-related GTP-binding protein RhoB
Gene Symbols RHOB
Host Species Alpaca
Immunogen Recombinant Human RHOB.
Purification Method Metal-chelate chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype VHH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.