missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOBTB2 Antibody (DBC2/3362), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08659FR
This item is not returnable.
View return policy
Description
RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| RHOBTB2 | |
| Monoclonal | |
| DyLight 680 | |
| DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2 | |
| Human recombinant RHOBTB2 protein fragment (around aa554-604) (exact sequence is proprietary) (Uniprot: Q9BYZ6) | |
| 100 μg | |
| Signal Transduction | |
| 23221 | |
| Store at 4°C in the dark. | |
| IgG2c λ |
| Immunohistochemistry (Paraffin), Protein Array | |
| DBC2/3362 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction