missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoD Antibody (OTI2F7), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73889FR
This item is not returnable.
View return policy
Description
RhoD Monoclonal specifically detects RhoD in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| RhoD | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RHOD | |
| Full length recombinant protein of human RhoD (NP_055393) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2F7 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| ARHDRHOM, ras homolog D, ras homolog gene family, member A, ras homolog gene family, member D, Rho, RhoD, RhoHP1, rho-related GTP-binding protein RhoD, Rho-related protein HP1 | |
| Mouse | |
| Affinity Purified | |
| mTOR Pathway, Signal Transduction | |
| 29984.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction