missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RhoD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101850
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RHOD.
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifications
| RhoD | |
| Polyclonal | |
| Unconjugated | |
| RHOD | |
| AI326383; aplysia ras-related homolog D; Arhd; LOW QUALITY PROTEIN: rho-related GTP-binding protein RhoD; ras homolog D; ras homolog D (RhoD); ras homolog family member D; ras homolog gene family, member A; ras homolog gene family, member D; Rh(o); Rh(o)ii; Rho; RHOD; RhoHP1; RHOM; Rho-related GTP-binding protein RhoD; Rho-related protein HP1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29984 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00212 | |
| RHOD | |
| A synthesized peptide derived from human RHOD(Accession O00212), corresponding to amino acid residues K121-C171. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction