Learn More
Invitrogen™ Rhodopsin Monoclonal Antibody (4D2), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545245
Description
1 μg/mL of MA5-45245 was sufficient for detection of rhodopsin in 10 μg of rat eye lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates.
Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
Specifications
| Rhodopsin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| P02699, P08100, P15409 | |
| RHO | |
| Bovine Rhodopsin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Bovine, Amphibian, Fish, Avian, Shark | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 4D2 | |
| FITC | |
| RHO | |
| CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho | |
| Mouse | |
| Protein G | |
| RUO | |
| 103176745, 212541, 509933, 6010 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.