missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rhodopsin Monoclonal Antibody (A531)

Mouse Monoclonal Antibody
€ 504.00 - € 1750.00
Specifications
| Antigen | Rhodopsin |
|---|---|
| Clone | A531 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17885257
|
Invitrogen™
MA547372 |
500 μL |
€ 1750.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17855127
![]() |
Invitrogen™
MA547471 |
100 μL |
€ 504.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhodopsin Monoclonal Antibody for Western Blot, ICC/IF, IHC
Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.Specifications
| Rhodopsin | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| O18766, P02699, P08100, P15409, P51489 | |
| 100056641, 212541, 24717, 397437, 509933, 6010 | |
| Purified bovine rhodopsin. | |
| Antibody |
| A531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| RHO | |
| CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho | |
| RHO | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title