missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoF Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98649AF647
This item is not returnable.
View return policy
Description
RhoF Polyclonal specifically detects RhoF in Human samples. It is validated for Western Blot.
Specifications
| RhoF | |
| Polyclonal | |
| Western Blot | |
| FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF | |
| Produced in rabbits immunized with E. coli-derived Human RhoF fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 54509 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction