missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98649-50ul
This item is not returnable.
View return policy
Description
RhoF Polyclonal antibody specifically detects RhoF in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| RhoF | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate | |
| FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF | |
| Produced in rabbits immunized with E. coli-derived Human RhoF fragment. | |
| 50 μg | |
| Signal Transduction | |
| 54509 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction