missing translation for 'onlineSavingsMsg'
Learn More

RhoGAP Antibody, Novus Biologicals™

Product Code. 18668386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18668386 25 μL 25µL
18234123 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18668386 Supplier Novus Biologicals Supplier No. NBP25862025ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RhoGAP Polyclonal specifically detects RhoGAP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoGAP
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias growth factor independent 2, p100 RasGAP-associated p105 protein, p105 RhoGAP, p190-BGFI2, p190BRhoGAP, Rho GTPase activating protein 5, rho GTPase-activating protein 5, RHOGAP5, Rho-type GTPase-activating protein 5
Gene Symbols ARHGAP5
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PLAHPEDMDPSDNYAEPIDTIFKQKGYSDEIYVVPDDSQNRIKIRNSFVNNTQGDEENGFSDRTSKSHGERRPSKYKYKSKTLFSKAKSYYRRTHSDASDDE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 394
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.