missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGDI Mouse anti-Human, Mouse, Rat, Clone: 1B1G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66480-1-IG-150UL
This item is not returnable.
View return policy
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.Specifications
| RhoGDI | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P52565, Q5XI73, Q99PT1 | |
| Arhgdia | |
| RhoGDI Fusion Protein Ag0791 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1B1G9 | |
| Unconjugated | |
| ARHGDIA | |
| aGDI, ARHGDIA, GDIA1, Rho GDI 1, Rho GDI alpha, RHOGDI, RHOGDI 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 192662, 360678, 396 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction