Learn More
RHOJ Monoclonal Antibody (OTI5C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505594
Encompass_Preferred
 View more versions of this product
Product Code. 17554252
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ARHJ belongs to the Rho family of small GTP-binding proteins. Rho proteins regulate the dynamic assembly of cytoskeletal components for several physiologic processes, such as cell proliferation and motility and the establishment of cell polarity. They are also involved in pathophysiologic process, such as cell transformation and metastasis.
Specifications
Specifications
RHOJ
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H4E5
RHOJ
Full length human recombit protein of human RHOJ produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI5C7
Unconjugated
RHOJ
ARHJ, RASL7B, TC10B, TCL
Mouse
Affinity Chromatography
RUO
57381
-20° C, Avoid Freeze/Thaw Cycles