Learn More
Rhot1 Antibody (CL1095) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52965FR
Product Code. 30303069
  • € 737.00 / 0.10mL
Description
Rhot1 Monoclonal antibody specifically detects Rhot1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rhot1
Monoclonal
DyLight 680
ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member T1
This antibody was developed using a recombinant protein derived from Q8IXI2, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
55288
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL1095
50mM Sodium Borate
Mouse
Protein A purified