missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rhot1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-27895
This item is not returnable.
View return policy
Description
Rhot1 Polyclonal antibody specifically detects Rhot1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Rhot1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member T1 | |
| Rhot1 antibody was raised against a 15 amino acid peptide near the amino terminus of human Rhot1. The immunogen is located within amino acids 40 - 90 of Rhot1. | |
| 0.1 mg | |
| Signal Transduction | |
| 55288 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Q8IXI2 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction