missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHOT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572835
This item is not returnable.
View return policy
Description
RHOT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.
Specifications
| RHOT1 | |
| Polyclonal | |
| Unconjugated | |
| RHOT1 | |
| 2210403N23Rik; AA415293; AF244542; AI834919; Arht1; C430039G08Rik; LOW QUALITY PROTEIN: mitochondrial Rho GTPase 1; MGC128572 protein; Miro1; MIRO-1; mitochondrial Rho (MIRO) GTPase 1; mitochondrial Rho GTPase 1; rac-GTP binding protein-like protein; Rac-GTP-binding protein-like protein; ras homolog family member T1; Ras homolog gene family member T1; ras homolog gene family, member T1; Rhot1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 303351, 55288, 59040 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8BG51, Q8IXI2 | |
| RHOT1 | |
| RHOT1 antibody for a 15 amino acid peptide near the amino terminus of human RHOT1. The immunogen is within amino acids 40 - 90 of RHOT1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction