missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rhox10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523135
This item is not returnable.
View return policy
Description
Immunogen sequence homology: human/rabbit (82%), canine (82%).
Reproductive homeobox on X chromosome 10, also known as Reproductive homeobox 10, is a recently identified nuclear protein product of a 12 X-linked novel homeobox gene family that plays an important role during postnatal testis development. The members of this family are characterized by a conserved 60-amino-acid DNA-binding homeodomain. These transcription factors not only play an important role in various developmental processes such as DNA-dependent transcriptional regulation, body-plan specification, pattern formation, and cell fate determination, but also in understanding the development and physiology of the human reproductive system.
Specifications
| Rhox10 | |
| Polyclonal | |
| Unconjugated | |
| Rhox10 | |
| reproductive homeobox 10; reproductive homeobox on chromosome X, 10; reproductive homeobox on X chromosome 10; Rhox10 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| Whole serum with 50% glycerol and 0.025% sodium azide | |
| 0 | |
| Rhox10 | |
| Within the range of amino acids 90-118 of mouse Rhox-10 protein. | |
| RUO | |
| 434769 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction