missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHOXF1 Monoclonal Antibody (4D12)
GREENER_CHOICE

Product Code. 15523192
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15523192 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15523192 Supplier Invitrogen™ Supplier No. MA523128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MARSLVHDTV FYCLSVYQVK ISPTPQLGAA SSAEGHVGQG APGLMGNMNP EGGVNHENGM NRDGGMIPEG GGGNQEPRQQ PQPPPEEPAQ AAMEGPQPEN MQPRTRRTKF.

This gene is a member of the PEPP subfamily of paired-like homoebox genes. The gene may be regulated by androgens and epigenetic mechanisms. The encoded nuclear protein is likely a transcription factor that may play a role in human reproduction. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D12
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RHOXF1
Gene Accession No. Q8NHV9
Gene Alias OTEX; ovary-, testis- and epididymis-expressed gene protein; Paired-like homeobox protein PEPP-1; PEPP subfamily gene 1; PEPP1; Rhox homeobox family member 1; Rhox homeobox family, member 1; RHOXF1; ZKSCAN21
Gene Symbols RHOXF1
Host Species Mouse
Immunogen RHOXF1 (NP_644811.1, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158800
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.