missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHOXF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15709850 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15709850 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15709850 Supplier Invitrogen™ Supplier No. PA551892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DQCSQYMTSL LSPAVDDEKE LQDMNAMVLS LTEEVKEEEE DAQPEPEQGT AAGEKLKSAG AQGGEEKDGG GEEKDGGGAG VPGHLWEGDL EGTSGSDGNV EDSDQSEKEP GQQ Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 27%.

This gene, which encodes a transcriptional repressor, is one of two paralogous X-linked homeobox-containing genes and is highly expressed in a variety of cancers. In addition, the encoded protein associates with the cell membrane and with microtubules, and is concentrated at the leading edge of migratory cells., Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOXF2
Gene Accession No. Q9BQY4
Gene Alias cancer/testis antigen 107; CT107; homeobox protein from AL590526; paired-like homeobox protein PEPP-2; PEPP subfamily gene 2; PEPP2; PEPP-2; Rhox homeobox family member 2; Rhox homeobox family, member 2; RHOXF2; Testis homeobox gene 1; THG1
Gene Symbols RHOXF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHOXF2. Recombinant protein control fragment (Product #RP-92207).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.