missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHPN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12671-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 77 kDa, and this antibody specifically recognizes the 77 kDa protein.
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.Specifications
| RHPN2 | |
| Polyclonal | |
| Unconjugated | |
| RHPN2 | |
| 76 kDa RhoB effector protein, GTP Rho binding protein 2, p76RBE, RhoBP, Rhophilin 2, RHPN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 308516, 52428, 85415 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BWR8, Q8IUC4 | |
| Rhpn2 | |
| RHPN2 Fusion Protein Ag3353 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction