Learn More
Abnova™ Ribonuclease A (Bovine), Molecular Biology Grade
Product Code. 16292240
Click to view available options
:
1mg
Product Code. 16292240
Brand: Abnova™ P5223.1mg
Used for Enzyme
#REF!
Specifications
Accession Number P61823
Concentration 5 mg/mL
For Use With (Application) Enzyme Activity
Formulation In 50% glycerol
Gene ID (Entrez) 282340
Molecular Weight (g/mol) 13.7kDa
Name Ribonuclease A (Bovine), Molecular Biology Grade
Preparation Method Native protein purified from Bovine Pancreas
Quantity 1 mg
Storage Requirements Store at 4°C. For long term storage store at -20°C.
Regulatory Status RUO
Gene Alias RNS1
Common Name RNASE1
Gene Symbol RNASE1
Assays Ribonuclease Assay; RNA removal during DNA isolation; RNA sequence analysis; RNase protection assays; RNA quantification or mapping.
Biological Activity ≥2,000 units per mg protein. One Kunitz unit is the amount of enzyme required to cause an increase in absorbance of 1.0 at 260 nm at 37°C (pH 5.0) when yeast rRNA is hydrolyzed to acid-soluble oligonucleotides. 50 units are equivalent to 1 Kunitz unit.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas
Form Liquid