missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease Inhibitor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99628-100ul
This item is not returnable.
View return policy
Description
Ribonuclease Inhibitor Polyclonal antibody specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Ribonuclease Inhibitor | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:3000 | |
| Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Ribonuclease Inhibitor. | |
| 100 μg | |
| metabolism, Proteases & Other Enzymes | |
| 6050 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction