missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribophorin I Antibody (OTI3H3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73898PCP
This item is not returnable.
View return policy
Description
Ribophorin I Monoclonal specifically detects Ribophorin I in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications
| Ribophorin I | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| RPN1 | |
| Full length recombinant protein of human Ribophorin I (NP_002941) produced in HEK293T cells. | |
| 0.1 mL | |
| 6184.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI3H3 | |
| Flow Cytometry | |
| DKFZp686B16177, dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, EC 2.4.1.119, oligosaccharyltransferase 1 homolog, OST1, ribophorin IRBPH1, ribophorin-1, RPN-I | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction