missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribophorin II Antibody (OTI1G9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73907F
This item is not returnable.
View return policy
Description
Ribophorin II Monoclonal specifically detects Ribophorin II in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Ribophorin II | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| RPN2 | |
| Full length recombinant protein of human Ribophorin II (NP_002942) produced in HEK293T cells. | |
| 0.1 mL | |
| 6185.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1G9 | |
| Immunohistochemistry | |
| RIBIIREC 2.4.1.119, ribophorin IIDolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit, Ribophorin-2, RPNII, RPN-IIdolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2, SWP1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction