missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein L24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15102
This item is not returnable.
View return policy
Description
Ribosomal Protein L24 Polyclonal specifically detects Ribosomal Protein L24 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Ribosomal Protein L24 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| RPL24 | |
| Recombinant protein encompassing a sequence within the center region of human RPL24. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| 60S ribosomal protein L24, 60S ribosomal protein L30, L24, ribosomal protein L30, RPL24 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6152 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction