Learn More
Invitrogen™ Ribosome-inactivating protein Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598352
Product Code. 16501182
  • € 464.00 / 100µg
Description
Ribosome-inactivating protein Polyclonal Antibody for Western Blot, ELISA
Has cytotoxic activity towards cancer cells, but not normal cells. Inhibits the growth of the human leukemia cell line K562, the murine melanoma cell line B16 and the lung adenocarcinoma cell line A549 with IC50 values of 88.1 nM, 63.4 nM and 359.3 nM respectively. [Uniprot].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ribosome-inactivating protein
Polyclonal
Unconjugated
cucurmosin; Ribosome-inactivating protein cucurmosin; rRNA N-glycosidase
Recombinant Cucurbita moschata Ribosome-inactivating protein cucurmosin protein (1-244AA).
100 μg
Primary
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
5 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Rabbit
Protein G
RUO
Plant