missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosome maturation protein SBDS Antibody (1E8), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22594AF594
This item is not returnable.
View return policy
Description
Ribosome maturation protein SBDS Monoclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Ribosome maturation protein SBDS | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SBDS | |
| Recombinant human SBDS (1-250aa) purified from E. coli | |
| metabolism | |
| 51119 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 100000000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction