missing translation for 'onlineSavingsMsg'
Learn More

RIC1 Antibody, Novus Biologicals™

Product Code. 18627535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18627535 25 μL 25µL
18633569 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18627535 Supplier Novus Biologicals Supplier No. NBP24901225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RIC1 Polyclonal antibody specifically detects RIC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIC1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias BA207C16.1, CIP150, Connexin 43-Interacting Protein 150 KDa, Connexin-43-Interacting Protein Of 150 KDa, Protein RIC1 Homolog, RAB6A GEF Complex Partner 1, RIC1, RIC1 Homolog, RAB6A GEF Complex Partner 1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: YCATLDGFAVVFNDGKVGFITPVSSRFTAEQLHGVWPQDVVDGTCVAVNNKYRLMAFGCVSGSVQVYTIDNSTGAMLLSHKLELTAKQYPD
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57589
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.