missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC8A Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36536H
This item is not returnable.
View return policy
Description
RIC8A Polyclonal antibody specifically detects RIC8A in Human, Mouse samples. It is validated for Western Blot
Specifications
| RIC8A | |
| Polyclonal | |
| Western Blot | |
| RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A | |
| The immunogen recognized by the antibody maps to a region between residue 418 to 468 of human Resistance to Inhibitors of Cholinesterase 8 Homolog A using the numbering given in entry EAW61245.1. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 60626 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction