missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC8A Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1473
This item is not returnable.
View return policy
Description
RIC8A Polyclonal specifically detects RIC8A in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| RIC8A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 60626 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_068751. | |
| RIC8A | |
| Peptide with sequence CLSSDPDSDPD corresponding to C-Terminus according to NP_068751.4. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction