missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC8A Antibody (OTI3G3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73908CL1
This item is not returnable.
View return policy
Description
RIC8A Monoclonal antibody specifically detects RIC8A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RIC8A | |
| Monoclonal | |
| CoraFluor 1 | |
| RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A | |
| Full length human recombinant protein of human RIC8A (NP_068751) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI3G3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 60626 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction