missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC8A Antibody (OTI3G3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73908UV
This item is not returnable.
View return policy
Description
RIC8A Monoclonal specifically detects RIC8A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RIC8A | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RIC8A | |
| Full length recombinant protein of human RIC8A (NP_068751) produced in HEK293T cells. | |
| 0.1 mL | |
| 60626.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?