missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIC8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116432
This item is not returnable.
View return policy
Description
RIC8B Polyclonal Antibody for Western Blot
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins by exchanging bound GDP for free GTP.
Specifications
| RIC8B | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6DRJ9 | |
| RIC8B | |
| KLH conjugated synthetic peptide between 444-478 amino acids from the C-terminal region of DANRE ric8b. | |
| 200 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RIC8B | |
| BC051080; brain synembrin; brain synembryn; fj97c10; heterotrimeric G protein guanine nucleotide exchange factor-like protein Ric-8B; hSyn; Protein Ric-8B; resistance to inhibitors of cholinesterase 8 homolog B; resistance to inhibitors of cholinesterase 8 homolog B (C. elegans); resistance to inhibitors of cholinesterase 8B; RIC8; Ric-8; RIC8 guanine nucleotide exchange factor B; ric8b; Ric-8b; synbl; synembryn like; synembryn-B; Synembryn-like; Unknown (protein for IMAGE:7408979); wu:fj97c10 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 334503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction