Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
1 gal.
Type:
Alcoholic Eosin Y Stain
Eosin Phloxine Stain
Unit Size:
3.8 L
Pack of 4
Description
- Strict quality control of raw materials and finished product ensures a high quality product every time
- Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
- Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
- Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
Specifications
Specifications
| Type | Eosin Phloxine Stain |
| Quantity | 1 pt. |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
- H225-Highly flammable liquid and vapour.
- H371-May cause damage to organs.
- EUH210-Safety data sheet available on request.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethyl alcohol, methyl alcohol, isopropylalcohol, acetic acid, eosin-Y dye, spiro[isobenzofuran-1(3H