missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RICS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38408-25ul
This item is not returnable.
View return policy
Description
RICS Polyclonal specifically detects RICS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RICS | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| A7KAX9 | |
| ARHGAP32 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AVATTEDNLSSSYSAVALDKAYFQTDRPAEQFHLQNNAPGNCDHPLPETTATGDPTHSNTTESGEQHHQVDLTGNQPHQAYLSGDPEKARITSVP | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| brain-specific Rho GTP-ase-activating protein, Brain-specific Rho GTPase-activating protein, GAB-associated CDC42, GAB-associated Cdc42/Rac GTPase-activating protein, GC-GAPGTPase regulator interacting with TrkA, GRITp200RhoGAP, KIAA0712p250GAP, MGC1892, rac GTPase activating protein, Rho GTPase activating protein 32, rho GTPase-activating protein 32, Rho/Cdc42/Rac GTPase-activating protein RICS, RhoGAP involved in the beta-catenin-N-cadherin and NMDA receptor signaling, RhoGAP involved in the -catenin-N-cadherin and NMDA receptor signaling, Rho-type GTPase-activating protein 32, RICSPX-RICS | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9743 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction