missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rictor Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-611C
This item is not returnable.
View return policy
Description
Rictor Polyclonal specifically detects Rictor in Human samples. It is validated for Western Blot.
Specifications
| Rictor | |
| Polyclonal | |
| Western Blot | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB100-611 is specific for human Rictor protein. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RICTOR | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Rapamycin-Insensitive Companion of mTOR using the numbering given in TrEMBL entry Q6R327 (GeneID 253260). | |
| 100 μL | |
| Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
| 253260 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction