missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rictor Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-612H
This item is not returnable.
View return policy
Description
Rictor Polyclonal antibody specifically detects Rictor in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rictor | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| The immunogen recognized by this antibody maps to a region between residue 1650 and the C-terminus (residue 1708) of human Rapamycin-Insensitive Companion of mTOR using the numbering given in TrEMBL entry Q6R327 (GeneID 253260). | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
| 253260 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction